Assay Details
Target Gene Details
Entrez Gene ID: | 21749 |
Gene Name: | telomeric repeat binding factor 1 |
Gene Aliases: |
Pin2, Trbf1, Trf1 |
Location: |
Chr.1:15804982-15844052 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Terf1 | NM_001286628.1 | 3 | 465 | NP_001273557.1 |
NM_009352.3 | 3 | 465 | NP_033378.1 | |
AB109563.1 | 3 | 445 | BAC76782.1 | |
AK021235.1 | 3 | 455 | BAB32340.1 | |
AK036615.1 | 3 | 468 | ||
AK132022.1 | 3 | 448 | BAE20947.1 | |
BB652245.1 | 3 | 466 | ||
BC034866.1 | 3 | 453 | AAH34866.1 | |
U65586.1 | 3 | 445 | AAB53970.1 |