Assay Details
Target Gene Details
Entrez Gene ID: | 20671 |
Gene Name: | SRY (sex determining region Y)-box 17 |
Gene Aliases: |
- |
Location: |
Chr.1:4490928-4497388 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sox17 | NM_001289464.1 | 1 | 925 | NP_001276393.1 |
NM_001289465.1 | 1 | 925 | NP_001276394.1 | |
NM_001289466.1 | 1 | 925 | NP_001276395.1 | |
NM_001289467.1 | 1 | 925 | NP_001276396.1 | |
NM_011441.5 | 1 | 925 | NP_035571.1 | |
AK004781.1 | BAB23559.1 | |||
AK076707.1 | 1 | 926 | ||
D49473.1 | 1 | 334 | BAA08443.1 |
More Information
Set Membership: |
Intragenic Exonic |