Assay Details
Target Gene Details
Entrez Gene ID: | 240690 |
Gene Name: | suppression of tumorigenicity 18 |
Gene Aliases: |
AV348974, Myt3, NZF-3, Nzf3, mKIAA0535 |
Location: |
Chr.1:6487231-6860940 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
St18 | NM_001244692.1 | 12 | 1905 | NP_001231621.1 |
NM_001244693.1 | 11 | 1827 | NP_001231622.1 | |
NM_173868.2 | 10 | 1718 | NP_776293.1 | |
NR_045188.1 | 12 | 1869 | ||
NR_045189.1 | 11 | 1791 | ||
AB097467.1 | 10 | 1743 | ||
AJ536155.1 | 12 | 1869 | CAD60089.1 | |
AK032557.1 | 12 | 1870 | ||
AK122303.1 | 10 | 1667 | BAC65585.1 | |
AK133743.1 | 12 | 1835 | BAE21817.1 | |
AK137440.1 | 10 | 1668 | BAE23351.1 | |
AK141513.1 | 10 | 1627 | BAE24709.1 | |
AK147320.1 | 12 | 1906 | BAE27844.1 | |
AK147664.1 | 12 | 1906 | BAE28057.1 | |
BC032273.1 | 5 | 632 | AAH32273.1 | |
BC118528.1 | 11 | 1741 |
More Information
Set Membership: |
Intragenic Exonic |