Assay Details
Target Gene Details
Entrez Gene ID: | 73122 |
Gene Name: | transforming growth factor, beta receptor associated protein 1 |
Gene Aliases: |
3110018K12Rik, AU024090, AU042859, AW742551, Trap1 |
Location: |
Chr.1:43047265-43098622 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tgfbrap1 | NM_001013025.2 | NP_001013043.1 | ||
AK141742.1 | BAE24818.1 | |||
BC138866.1 | ||||
BC138867.1 |