Assay Details
Target Gene Details
Entrez Gene ID: | 21808 |
Gene Name: | transforming growth factor, beta 2 |
Gene Aliases: |
BB105277, Tgf-beta2, Tgfb-2 |
Location: |
Chr.1:186622787-186709033 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tgfb2 | NM_009367.3 | 2 | 1640 | NP_033393.2 |
AK141552.1 | 3 | 1022 | ||
AK159690.1 | 2 | 204 | BAE35291.1 | |
BC011170.1 | 2 | 548 | AAH11170.1 | |
X57413.1 | 2 | 1639 | CAA40672.1 |
More Information
Set Membership: |
Intragenic Exonic |