Assay Details
Target Gene Details
Entrez Gene ID: | 54380 |
Gene Name: | SWI/SNF related matrix associated, actin dependent regulator of chromatin, subfamily a-like 1 |
Gene Aliases: |
6030401P21Rik, AI851716, Mharp |
Location: |
Chr.1:72536687-72636790 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smarcal1 | NM_018817.2 | 14 | 2490 | NP_061287.2 |
AF088884.1 | 14 | 2470 | AAF24985.1 | |
AK043129.1 | 3 | 305 | BAC31469.1 | |
AK077878.1 | 14 | 2468 | BAC37047.1 | |
AK083488.1 | 14 | 2468 | BAC38933.1 | |
AK146767.1 | 14 | 2486 | ||
AK154020.1 | 14 | 2473 | BAE32320.1 | |
AK161570.1 | 13 | 3782 | ||
BC029078.1 | 14 | 2425 | AAH29078.1 |
More Information
Set Membership: |
Intragenic Exonic |