Assay Details
Target Gene Details
Entrez Gene ID: | 320718 |
Gene Name: | solute carrier family 26, member 9 |
Gene Aliases: |
E030002L01Rik |
Location: |
Chr.1:131744022-131771504 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc26a9 | NM_177243.4 | 5 | 572 | NP_796217.2 |
AK078966.1 | 5 | 573 | BAC37482.1 | |
AK086815.1 | 5 | 573 | BAC39749.1 | |
AY034145.1 | 4 | 485 | AAK54448.1 |
More Information
Set Membership: |
Intragenic Exonic |