Assay Details
Target Gene Details
Entrez Gene ID: | 19731 |
Gene Name: | ral guanine nucleotide dissociation stimulator,-like 1 |
Gene Aliases: |
Rgl, mKIAA0959 |
Location: |
Chr.1:152516768-152766351 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rgl1 | NM_016846.3 | NP_058542.2 | ||
AK144218.1 | BAE25780.1 | |||
AK169109.1 | BAE40891.1 | |||
AK169180.1 | BAE40958.1 | |||
AK173071.1 | BAD32349.1 | |||
BC017678.1 | AAH17678.1 | |||
U14103.1 | AAA64950.1 |
More Information
Set Membership: |
Intragenic Non-exonic |