Assay Details
Target Gene Details
Entrez Gene ID: | 226971 |
Gene Name: | pleckstrin homology domain containing, family B (evectins) member 2 |
Gene Aliases: |
2310009M15Rik, AU018239, Evt2, Phdc, evt-2 |
Location: |
Chr.1:34849959-34879585 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plekhb2 | NM_145516.2 | 7 | 591 | NP_663491.1 |
NM_175421.1 | 7 | 591 | NP_780630.1 | |
AF189817.1 | 7 | 551 | AAF01332.1 | |
AK039771.1 | 7 | 559 | BAC30448.1 | |
AK082118.1 | 7 | 544 | BAC38413.1 | |
AK148653.1 | 7 | 574 | BAE28632.1 | |
AK149868.1 | 7 | 551 | BAE29136.1 | |
AK150074.1 | 7 | 551 | BAE29285.1 | |
AK166091.1 | 7 | 559 | BAE38565.1 | |
BC057994.1 | 7 | 559 | AAH57994.1 | |
CK625615.1 |