Assay Details
Target Gene Details
Entrez Gene ID: | 18802 |
Gene Name: | phospholipase C, delta 4 |
Gene Aliases: |
4921507K24Rik |
Location: |
Chr.1:74541972-74565977 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plcd4 | NM_001081456.1 | NP_001074925.1 | ||
AK039149.1 | BAC30256.1 | |||
AK140231.1 | BAE24292.1 |
Target Gene Details
Entrez Gene ID: | 319651 |
Gene Name: | ubiquitin specific peptidase 37 |
Gene Aliases: |
4932415L06Rik, C330008N13Rik |
Location: |
Chr.1:74435510-74544286 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Usp37 | NM_176972.4 | NP_795946.2 | ||
AK030013.1 | BAC26734.1 | |||
AK133317.1 | 1 | 1360 | ||
BC139091.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |