Assay Details
Target Gene Details
Entrez Gene ID: | 65102 |
Gene Name: | Ngg1 interacting factor 3-like 1 (S. pombe) |
Gene Aliases: |
1110030G24Rik |
Location: |
Chr.1:58445156-58462276 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nif3l1 | AF284439.1 | 1 | 256 | AAG45961.1 |
AK131631.1 | 1 | 142 | BAE20729.1 | |
AK158616.1 | 1 | 162 | BAE34583.1 |
Target Gene Details
Entrez Gene ID: | 70225 |
Gene Name: | peptidylprolyl isomerase (cyclophilin)-like 3 |
Gene Aliases: |
2310076N22Rik, 2510026K04Rik, Cyp10l |
Location: |
Chr.1:58430993-58445527 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppil3 | NM_001285826.1 | 1 | 81 | NP_001272755.1 |
NM_001285827.1 | 1 | 81 | NP_001272756.1 | |
NM_027351.3 | 1 | 81 | NP_081627.1 | |
NM_027374.3 | 1 | 81 | NP_081650.2 | |
AF315803.1 | AAO32944.1 | |||
AK010207.1 | 1 | 85 | BAB26768.1 | |
BB844274.1 | 1 | 63 | ||
BC060993.1 | 1 | 49 | AAH60993.1 | |
BC061645.1 | AAH61645.1 | |||
BY288296.1 | ||||
CJ166559.1 | ||||
CJ170065.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |