Assay Details
Target Gene Details
Entrez Gene ID: | 68507 |
Gene Name: | protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein (liprin), alpha 4 |
Gene Aliases: |
1110008G13Rik, AI448359, AI852265, C81506 |
Location: |
Chr.1:134296783-134344750 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppfia4 | NM_001144855.1 | 25 | 3575 | NP_001138327.1 |
AK003571.1 | 4 | 433 | ||
AK078263.1 | 2 | 612 | ||
AK170782.1 | 3 | 468 | ||
AK173053.1 | 24 | 3038 | BAD32331.1 | |
EU568872.1 | 25 | 3575 |