Assay Details
Target Gene Details
Entrez Gene ID: | 19258 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 4 |
Gene Aliases: |
PTPMEG, Ptn4, TEP, TEP/mPTPMEG, hPTP-MEG |
Location: |
Chr.1:119652467-119837542 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptpn4 | NM_019933.2 | 1 | 41 | NP_064317.2 |
AK046683.1 | 1 | 317 | BAC32836.1 | |
BE985496.1 | 1 | 41 |
More Information
Set Membership: |
Intragenic Exonic |