Assay Details
Target Gene Details
Entrez Gene ID: | 213527 |
Gene Name: | parathyroid hormone 2 receptor |
Gene Aliases: |
Pthr2 |
Location: |
Chr.1:65280282-65389625 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pth2r | NM_139270.2 | NP_644676.1 | ||
AF332077.1 | AAK56105.1 | |||
AF332078.1 | AAK56106.1 | |||
AK045576.1 | BAC32420.1 |
More Information
Set Membership: |
Intragenic Non-exonic |