Assay Details
Target Gene Details
Entrez Gene ID: | 227102 |
Gene Name: | ORM1-like 1 (S. cerevisiae) |
Gene Aliases: |
C730042F17Rik |
Location: |
Chr.1:53297095-53310245 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ormdl1 | NM_145517.4 | NP_663492.3 | ||
AK050385.1 | BAC34226.1 | |||
AK052927.1 | ||||
AK077562.1 | BAC36865.1 | |||
BC012315.1 | AAH12315.1 | |||
BC023695.1 | AAH23695.1 | |||
BC025572.1 | AAH25572.1 | |||
CN705165.1 |