Assay Details
Target Gene Details
Entrez Gene ID: | 171567 |
Gene Name: | NME/NM23 family member 7 |
Gene Aliases: |
D530024H21Rik, Nm23-M7, Nm23-r7 |
Location: |
Chr.1:164307662-164437300 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nme7 | NM_001293313.1 | 5 | 500 | NP_001280242.1 |
NM_138314.4 | 5 | 503 | NP_612187.2 | |
NM_178071.6 | 5 | 500 | NP_835172.1 | |
AF202048.1 | 4 | 404 | AAF20906.1 | |
AK043515.1 | 5 | 487 | ||
AK044962.1 | 5 | 474 | ||
AK085222.1 | 5 | 448 | BAC39392.1 | |
AK144915.1 | 5 | 504 | BAE26132.1 | |
AK164652.1 | 5 | 448 | BAE37861.1 | |
BC038021.1 | 6 | 530 | ||
BC049225.1 | 4 | 431 | AAH49225.1 |
More Information
Set Membership: |
Intragenic Exonic |