Assay Details
Target Gene Details
Entrez Gene ID: | 27392 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis, class N |
Gene Aliases: |
Gm20308, PIG-N |
Location: |
Chr.1:105518422-105663722 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pign | NM_013784.3 | NP_038812.2 | ||
AB030279.1 | BAA82619.1 | |||
AB030280.1 | BAA82620.1 | |||
BC021148.1 | AAH21148.1 |
More Information
Set Membership: |
Intragenic Non-exonic |