Assay Details
Target Gene Details
Entrez Gene ID: | 17936 |
Gene Name: | Ngfi-A binding protein 1 |
Gene Aliases: |
- |
Location: |
Chr.1:52455849-52500448 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nab1 | NM_008667.3 | NP_032693.2 | ||
AB221677.1 | ||||
BC005627.1 | AAH05627.1 | |||
BC016886.1 | AAH16886.1 | |||
U47008.1 | AAC52649.1 |