Assay Details
Target Gene Details
Entrez Gene ID: | 107895 |
Gene Name: | mannoside acetylglucosaminyltransferase 5 |
Gene Aliases: |
4930471A21Rik, 5330407H02Rik, AI480971, GlcNAc-TV |
Location: |
Chr.1:127204977-127488334 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mgat5 | NM_145128.3 | NP_660110.2 | ||
AF474154.1 | AAL83249.1 | |||
AF474155.1 | AAL83250.1 | |||
AK030402.1 | BAC26946.1 | |||
BC125533.1 | ||||
BC125535.1 |
More Information
Set Membership: |
Intragenic Non-exonic |