Assay Details
Target Gene Details
Entrez Gene ID: | 171567 |
Gene Name: | NME/NM23 family member 7 |
Gene Aliases: |
D530024H21Rik, Nm23-M7, Nm23-r7 |
Location: |
Chr.1:164307662-164437300 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nme7 | NM_001293313.1 | 13 | 1398 | NP_001280242.1 |
NM_138314.4 | 13 | 1401 | NP_612187.2 | |
AF202048.1 | 12 | 1302 | AAF20906.1 | |
AK144915.1 | 13 | 1402 | BAE26132.1 | |
AK160566.1 | 5 | 357 | ||
BC038021.1 | 14 | 1428 | ||
BC049225.1 | 12 | 1329 | AAH49225.1 | |
BU609451.1 | 2 | 341 | ||
CD775820.1 | 2 | 352 |
More Information
Set Membership: |
Intragenic Exonic |