Assay Details
Target Gene Details
Entrez Gene ID: | 17974 |
Gene Name: | non-catalytic region of tyrosine kinase adaptor protein 2 |
Gene Aliases: |
4833426I10Rik, Grb4, NCKbeta, Nck-2 |
Location: |
Chr.1:43445749-43570518 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nck2 | NM_010879.3 | NP_035009.3 | ||
AF043260.1 | AAC06353.1 | |||
AK014949.1 | ||||
AK051665.1 | BAC34709.1 | |||
BC034255.1 | AAH34255.1 |
More Information
Set Membership: |
Intragenic Non-exonic |