Assay Details
Target Gene Details
Entrez Gene ID: | 68421 |
Gene Name: | LMBR1 domain containing 1 |
Gene Aliases: |
0910001K20Rik, AV347960 |
Location: |
Chr.1:24678544-24766301 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lmbrd1 | NM_026719.2 | 2 | 320 | NP_080995.2 |
AK152170.1 | 2 | 131 | BAE31002.1 | |
AK153237.1 | 2 | 131 | BAE31829.1 | |
AK158982.1 | 2 | 156 | BAE34756.1 | |
AK167080.1 | 2 | 139 | BAE39240.1 | |
AY061989.1 | 1 | 31 | AAL38052.1 | |
BC025579.1 | 2 | 152 | AAH25579.1 | |
BC027774.1 | 2 | 203 | AAH27774.1 | |
BC031902.1 | 2 | 143 | AAH31902.1 | |
BC039053.1 | 2 | 234 | AAH39053.1 | |
BY291845.1 | 2 | 287 | ||
BY322390.1 |
More Information
Set Membership: |
Intragenic Exonic |