Assay Details
Target Gene Details
Entrez Gene ID: | 226922 |
Gene Name: | potassium voltage-gated channel, subfamily Q, member 5 |
Gene Aliases: |
7730402H11, 9230107O05Rik, AA589396, D1Mgi1 |
Location: |
Chr.1:21398399-21962650 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kcnq5 | NM_001160139.1 | NP_001153611.1 | ||
NM_023872.3 | NP_076361.1 | |||
AF263836.1 | AAF73447.1 | |||
AK033079.1 | BAC28145.1 | |||
AK147264.1 | ||||
AY679158.1 | AAT76442.1 |
More Information
Set Membership: |
Intragenic Non-exonic |