Assay Details
Target Gene Details
Entrez Gene ID: | 17082 |
Gene Name: | interleukin 1 receptor-like 1 |
Gene Aliases: |
DER4, Fit-1, Ly84, ST2L, St2, St2-rs1, T1, T1/ST2 |
Location: |
Chr.1:40404864-40465418 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Il1rl1 | NM_001025602.3 | NP_001020773.1 | ||
NM_001294171.1 | NP_001281100.1 | |||
NM_010743.3 | NP_034873.2 | |||
AK033417.1 | BAC28277.1 | |||
AK145118.1 | BAE26245.1 | |||
BC051445.1 | AAH51445.1 | |||
D13695.1 | BAA02854.1 | |||
M24843.1 | AAA40160.1 | |||
Y07519.1 | CAA68812.1 |
More Information
Set Membership: |
Intragenic Non-exonic |