Assay Details
Target Gene Details
Entrez Gene ID: | 16182 |
Gene Name: | interleukin 18 receptor 1 |
Gene Aliases: |
Il18ralpha, Il1rrp |
Location: |
Chr.1:40465552-40500854 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Il18r1 | NM_001161842.1 | 6 | 1000 | NP_001155314.1 |
NM_001161843.1 | 6 | 872 | NP_001155315.1 | |
NM_008365.2 | 5 | 925 | NP_032391.1 | |
AK087129.1 | 5 | 874 | ||
AK089237.1 | 6 | 1001 | BAC40807.1 | |
AK170705.1 | 6 | 961 | BAE41966.1 | |
BC023240.1 | 6 | 841 | AAH23240.1 | |
U43673.1 | 5 | 925 | AAC52437.1 |
More Information
Set Membership: |
Intragenic Exonic |