Assay Details
Target Gene Details
Entrez Gene ID: | 16153 |
Gene Name: | interleukin 10 |
Gene Aliases: |
CSIF, Il-10 |
Location: |
Chr.1:131019845-131024970 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Il10 | NM_010548.2 | NP_034678.1 | ||
AK152344.1 | BAE31139.1 | |||
BC120612.1 | ||||
BC137844.1 | ||||
M37897.1 | AAA39274.1 |