Assay Details
Target Gene Details
Entrez Gene ID: | 140559 |
Gene Name: | immunoglobulin superfamily, member 8 |
Gene Aliases: |
AA033172, AU041109, ESTM34, EWI-2, KCT-4, PGRL, R75384 |
Location: |
Chr.1:172312407-172319841 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Igsf8 | NM_080419.1 | NP_536344.1 | ||
AF411055.1 | AAL02217.1 | |||
AF439263.1 | AAN63626.1 | |||
AK140254.1 | ||||
AK146104.1 | BAE26904.1 | |||
AK146221.1 | BAE26989.1 | |||
AK150239.1 | BAE29403.1 | |||
AK158522.1 | BAE34544.1 | |||
AK168363.1 | BAE40298.1 | |||
AK206992.1 | ||||
AK213338.1 | ||||
AK214657.1 | ||||
BC048387.1 | AAH48387.1 |
More Information
Set Membership: |
Intragenic Non-exonic |