Assay Details
Target Gene Details
Entrez Gene ID: | 15926 |
Gene Name: | isocitrate dehydrogenase 1 (NADP+), soluble |
Gene Aliases: |
AI314845, AI788952, E030024J03Rik, Id-1, Idh-1, Idpc |
Location: |
Chr.1:65158616-65186479 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Idh1 | NM_001111320.1 | 1 | 84 | NP_001104790.1 |
BY787507.1 | 1 | 85 | ||
CF950262.1 |
More Information
Set Membership: |
Intragenic Exonic |