Assay Details
Target Gene Details
Entrez Gene ID: | 98376 |
Gene Name: | golgin, RAB6-interacting |
Gene Aliases: |
AI467484, NTKL-BP1, Scyl1bp1 |
Location: |
Chr.1:163384906-163403908 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gorab | NM_178883.5 | 1 | 63 | NP_849214.2 |
AK039170.1 | 1 | 70 | BAC30263.1 | |
AK043953.1 | 1 | 64 | BAC31712.1 | |
AK165909.1 | BAE38454.1 | |||
AK169151.1 | 1 | 56 | BAE40930.1 | |
AK170455.1 | 1 | 80 | ||
BC119132.1 | 1 | 46 | ||
BC119134.1 | 1 | 46 |