Assay Details
Target Gene Details
Entrez Gene ID: | 98752 |
Gene Name: | Fc receptor-like A |
Gene Aliases: |
BB219290, FCRL, FCRL1, FREB, Fcrlm1, Fcrx, Freb1, mFREB, mFcrX |
Location: |
Chr.1:170917594-170927590 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fcrla | NM_001160215.1 | 2 | 221 | NP_001153687.1 |
NM_145141.2 | 2 | 218 | NP_660123.1 | |
NR_027666.1 | 2 | 287 | ||
AF329487.1 | 2 | 185 | AAL23897.1 | |
AF426462.1 | 2 | 178 | AAL58112.1 | |
AF531424.1 | 2 | 207 | AAM97592.1 | |
AK041572.1 | 2 | 198 | BAC30990.1 | |
AK172370.1 | 2 | 211 | BAE42968.1 | |
BB637253.1 | 2 | 214 | ||
BC064708.1 | 2 | 221 | AAH64708.1 | |
CJ080700.1 | 2 | 260 |
More Information
Set Membership: |
Intragenic Exonic |