Assay Details
Target Gene Details
Entrez Gene ID: | 14645 |
Gene Name: | glutamate-ammonia ligase (glutamine synthetase) |
Gene Aliases: |
GS, Glns |
Location: |
Chr.1:153899929-153909723 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Glul | NM_008131.4 | NP_032157.2 | ||
AK137969.1 | ||||
AK159106.1 | BAE34822.1 | |||
AK160670.1 | BAE35950.1 | |||
AK162685.1 | BAE37022.1 | |||
AK168493.1 | BAE40380.1 | |||
AY044241.1 | AAK95328.1 | |||
BC015086.1 | AAH15086.1 | |||
X16314.1 | CAA34381.1 |
More Information
Set Membership: |
Intragenic Non-exonic |