Assay Details
Target Gene Details
Entrez Gene ID: | 227331 |
Gene Name: | GRB10 interacting GYF protein 2 |
Gene Aliases: |
2610016F01Rik, A830080H02Rik, AI852361, AW259676, BC006835, Tnrc15, mKIAA0642 |
Location: |
Chr.1:87326981-87450810 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gigyf2 | NM_001110212.2 | 10 | 1057 | NP_001103682.1 |
NM_146112.4 | 10 | 1075 | NP_666224.3 | |
AK079246.1 | 10 | 1027 | BAC37586.1 | |
AK122337.1 | 1 | 146 | BAC65619.1 | |
AK144058.1 | 10 | 1037 | BAE25675.1 | |
AK157733.1 | 1 | 2149 | ||
AK160878.1 | 8 | 869 | BAE36066.1 | |
AY176043.1 | 10 | 1021 | AAO46887.1 | |
BC089036.1 | 10 | 1031 |