Assay Details
Target Gene Details
Entrez Gene ID: | 226564 |
Gene Name: | flavin containing monooxygenase 4 |
Gene Aliases: |
D1Ertd532e |
Location: |
Chr.1:162793188-162815869 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fmo4 | NM_144878.1 | 3 | 293 | NP_659127.1 |
AF461145.1 | 3 | 293 | AAL66366.1 | |
AK146562.1 | 4 | 495 | ||
BC106858.1 | 2 | 284 | AAI06859.1 | |
BC106859.1 | 2 | 284 | AAI06860.1 |
More Information
Set Membership: |
Intragenic Exonic |