Assay Details
Target Gene Details
Entrez Gene ID: | 55990 |
Gene Name: | flavin containing monooxygenase 2 |
Gene Aliases: |
2310008D08Rik, 2310042I22Rik, AW107733 |
Location: |
Chr.1:162874321-162898712 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fmo2 | NM_018881.3 | NP_061369.2 | ||
AF184981.1 | 1 | 132 | AAD56413.1 | |
AK009224.1 | ||||
BC031415.1 | AAH31415.1 |
More Information
Set Membership: |
Intragenic Non-exonic |