Assay Details
Target Gene Details
Entrez Gene ID: | 213452 |
Gene Name: | dual serine/threonine and tyrosine protein kinase |
Gene Aliases: |
A930019K20Rik, C430014H23Rik, C820013G01, Ripk5 |
Location: |
Chr.1:132417181-132466959 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dstyk | NM_172516.4 | 6 | 1907 | NP_766104.2 |
AK043199.1 | 1 | 776 | BAC31487.1 | |
AK050542.1 | 2 | 153 | BAC34316.1 | |
AK083696.1 | 6 | 1805 | ||
AK132119.1 | 6 | 1782 | ||
AK145449.1 | 6 | 1752 | BAE26444.1 | |
AK172954.1 | 6 | 1753 | BAD32232.1 | |
AK183448.1 | 2 | 192 | ||
AK184349.1 | 2 | 192 | ||
AK195351.1 | ||||
AY208852.1 | 6 | 1754 | AAP42420.1 | |
AY429676.1 | 6 | 1793 | AAS55392.1 | |
BC089380.1 | 3 | 324 | AAH89380.1 | |
BC130226.1 |
More Information
Set Membership: |
Intragenic Exonic |