Assay Details
Target Gene Details
Entrez Gene ID: | 226982 |
Gene Name: | eukaryotic translation initiation factor 5B |
Gene Aliases: |
A030003E17Rik, BC018347, IF2 |
Location: |
Chr.1:37998000-38055579 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 17 - Intron 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif5b | NM_198303.2 | NP_938045.2 | ||
AK037164.1 | BAC29728.1 | |||
AK173016.1 | BAD32294.1 | |||
BC055393.1 | AAH55393.1 |
More Information
Set Membership: |
Intragenic Non-exonic |