Assay Details
Target Gene Details
Entrez Gene ID: | 13518 |
Gene Name: | dystonin |
Gene Aliases: |
2310001O04Rik, A830042E19Rik, AW554249, BP230, BPAG1-n, Bpag, Bpag1, Macf2, ah, athetoid, dt, mKIAA0728, nmf203, nmf339 |
Location: |
Chr.1:33908208-34308662 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dst | NM_001276764.1 | 23 | 3068 | NP_001263693.1 |
NM_010081.2 | 10 | 1509 | NP_034211.2 | |
NM_133833.3 | 20 | 2487 | NP_598594.2 | |
NM_134448.3 | 20 | 2487 | NP_604443.2 | |
AF396877.1 | 10 | 1500 | AAK83382.1 | |
AF396878.1 | 20 | 2499 | AAK83383.1 | |
AF396879.1 | 20 | 2499 | AAK83384.1 |
More Information
Set Membership: |
Intragenic Exonic |