Assay Details
Target Gene Details
Entrez Gene ID: | 13211 |
Gene Name: | DEAH (Asp-Glu-Ala-His) box polypeptide 9 |
Gene Aliases: |
AI326842, Ddx9, HEL-5, NDHII, RHA, mHEL-5 |
Location: |
Chr.1:153455756-153487686 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dhx9 | NM_007842.2 | NP_031868.2 | ||
AK135603.1 | ||||
AK141824.1 | BAE24846.1 | |||
AK148423.1 | ||||
AY512925.1 | AAR87796.1 | |||
BC089159.1 | AAH89159.1 | |||
U91922.1 | AAC05725.1 | |||
U92080.1 | AAB72087.1 |
More Information
Set Membership: |
Intragenic Non-exonic |