Assay Details
Target Gene Details
Entrez Gene ID: | 23836 |
Gene Name: | cadherin 20 |
Gene Aliases: |
Cdh7 |
Location: |
Chr.1:104768819-104995481 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdh20 | NM_011800.4 | 6 | 1101 | NP_035930.1 |
AF007116.1 | 6 | 1391 | AAD01278.1 | |
AK034475.1 | 6 | 1102 | BAC28721.1 | |
BC119606.2 | 6 | 1080 | ||
BC119607.2 | 6 | 1080 |
More Information
Set Membership: |
Intragenic Exonic |