Assay Details
Target Gene Details
Entrez Gene ID: | 67426 |
Gene Name: | aarF domain containing kinase 3 |
Gene Aliases: |
4632432J16Rik, AI462003, Cabc1, mKIAA0451 |
Location: |
Chr.1:180165238-180196020 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adck3 | NM_001163290.1 | NP_001156762.1 | ||
NM_023341.3 | NP_075830.2 | |||
AJ278735.1 | CAC00538.1 | |||
AK004791.1 | BAB23567.2 | |||
AK014605.2 | BAB29459.2 | |||
AK179829.1 | ||||
AK179958.1 | ||||
AK181234.1 | ||||
AK182957.1 | ||||
AK188107.1 | ||||
BC030937.1 | AAH30937.1 | |||
BY261815.1 | ||||
BY275962.1 |