Assay Details
Target Gene Details
Entrez Gene ID: | 12269 |
Gene Name: | complement component 4 binding protein |
Gene Aliases: |
AI195242, C4bpa |
Location: |
Chr.1:130635921-130661618 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C4bp | NM_007576.3 | NP_031602.3 | ||
AB094486.1 | BAC75872.1 | |||
AK136735.1 | BAE23113.1 | |||
AK149468.1 | BAE28899.1 | |||
AK149477.1 | BAE28904.1 | |||
AK162345.1 | ||||
BC012257.1 | AAH12257.1 | |||
M17122.1 | AAA37312.1 |