Assay Details
Target Gene Details
Entrez Gene ID: | 70456 |
Gene Name: | mitochondrial pyruvate carrier 2 |
Gene Aliases: |
0610006C01Rik, 2010002I07Rik, 2610205H19Rik, AA108335, Brp44, ESTM43 |
Location: |
Chr.1:165461208-165481214 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mpc2 | NM_027430.2 | NP_081706.1 | ||
AK002233.2 | BAC24983.1 | |||
AK011889.1 | BAB27898.1 | |||
AK151522.1 | BAE30471.1 | |||
BC018324.1 | AAH18324.1 |