Assay Details
Target Gene Details
Entrez Gene ID: | 210933 |
Gene Name: | adhesion G protein-coupled receptor B3 |
Gene Aliases: |
A830096D10Rik, Bai3 |
Location: |
Chr.1:25067476-25830206 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adgrb3 | NM_175642.4 | 13 | 2450 | NP_783573.4 |
AY168406.1 | 12 | 2189 | AAO27431.1 | |
AY255544.1 | AAO85056.1 | |||
BC032251.1 | 7 | 766 | AAH32251.1 | |
BC079670.1 | 12 | 1568 | AAH79670.1 | |
BC099951.1 | 13 | 2448 | AAH99951.1 |
More Information
Set Membership: |
Intragenic Exonic |