Assay Details
Target Gene Details
Entrez Gene ID: | 210933 |
Gene Name: | adhesion G protein-coupled receptor B3 |
Gene Aliases: |
A830096D10Rik, Bai3 |
Location: |
Chr.1:25067476-25830206 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adgrb3 | NM_175642.4 | 10 | 2096 | NP_783573.4 |
AK129162.1 | 9 | 1422 | BAC97972.1 | |
AY168406.1 | 9 | 1835 | AAO27431.1 | |
BC032251.1 | 4 | 412 | AAH32251.1 | |
BC079670.1 | 9 | 1214 | AAH79670.1 | |
BC099951.1 | 10 | 2094 | AAH99951.1 |
More Information
Set Membership: |
Intragenic Exonic |