Assay Details
Target Gene Details
Entrez Gene ID: | 71872 |
Gene Name: | aldehyde oxidase 4 |
Gene Aliases: |
2310003G12Rik, Aoh2 |
Location: |
Chr.1:58210397-58268600 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aox4 | NM_023631.2 | 9 | 964 | NP_076120.2 |
AF233581.1 | 9 | 891 | AAF98385.1 | |
AK158427.1 | 9 | 965 | BAE34503.1 | |
AK160953.1 | 9 | 930 | ||
BC117975.1 | 9 | 891 | ||
BC117976.1 | 9 | 891 |