Assay Details
Target Gene Details
Entrez Gene ID: | 23797 |
Gene Name: | thymoma viral proto-oncogene 3 |
Gene Aliases: |
AI851531, D930002M15Rik, Nmf350 |
Location: |
Chr.1:177020076-177258289 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Akt3 | NM_011785.3 | 4 | 366 | NP_035915.3 |
AF124142.1 | 4 | 356 | AAD29090.1 | |
BC066861.1 | 4 | 366 | AAH66861.1 |
More Information
Set Membership: |
Intragenic Exonic |