Assay Details
Target Gene Details
Entrez Gene ID: | 71724 |
Gene Name: | aldehyde oxidase 3 |
Gene Aliases: |
1200011D03Rik, AI326299, Aoh1, Aoh2 |
Location: |
Chr.1:58113136-58200452 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aox3 | NM_023617.2 | 4 | 386 | NP_076106.2 |
AF172276.2 | 4 | 462 | AAD51028.2 | |
AK054065.1 | 4 | 345 | BAC35643.1 | |
AK143048.1 | 4 | 725 | ||
AK149434.1 | 4 | 347 | ||
AY347569.1 | 4 | 263 | AAQ24537.1 | |
AY347570.1 | 4 | 263 | AAQ24538.1 | |
BC138876.1 | 4 | 365 |