Assay Details
Target Gene Details
Entrez Gene ID: | 11761 |
Gene Name: | aldehyde oxidase 1 |
Gene Aliases: |
AI196512, AI255253, Ao, Aox-1, Aox-2, Aox2, Moro, Ro |
Location: |
Chr.1:58029809-58106410 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aox1 | NM_009676.2 | 4 | 301 | NP_033806.2 |
AB017482.1 | 4 | 367 | BAA36834.1 | |
AF076216.1 | 4 | 266 | AAC99382.1 | |
AK137697.1 | 4 | 308 | BAE23466.1 | |
BC026132.1 | 4 | 293 | AAH26132.1 |