Assay Details
Target Gene Details
Entrez Gene ID: | 227290 |
Gene Name: | angio-associated migratory protein |
Gene Aliases: |
AU040907, Aamp-rs |
Location: |
Chr.1:74279840-74284738 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aamp | NM_001190444.1 | NP_001177373.1 | ||
NM_146110.3 | NP_666222.2 | |||
AK146470.1 | BAE27196.1 | |||
AK151820.1 | BAE30717.1 | |||
AK162750.1 | BAE37048.1 | |||
AK167621.1 | BAE39673.1 | |||
AK180202.1 | ||||
AK183212.1 | ||||
AK189765.1 | ||||
AK200115.1 | ||||
BC031795.1 | AAH31795.1 |