Assay Details
Target Gene Details
Entrez Gene ID: | 227290 |
Gene Name: | angio-associated migratory protein |
Gene Aliases: |
AU040907, Aamp-rs |
Location: |
Chr.1:74279840-74284738 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aamp | NM_001190444.1 | 1 | 158 | NP_001177373.1 |
NM_146110.3 | 1 | 158 | NP_666222.2 | |
AK146470.1 | 1 | 101 | BAE27196.1 | |
AK151820.1 | 1 | 70 | BAE30717.1 | |
AK162750.1 | 1 | 86 | BAE37048.1 | |
AK167621.1 | 1 | 82 | BAE39673.1 | |
BY101182.1 | 1 | 158 |
Target Gene Details
Entrez Gene ID: | 56695 |
Gene Name: | paroxysmal nonkinesiogenic dyskinesia |
Gene Aliases: |
2210013N15Rik, 2810403H05Rik, AI854243, Brp17, MNCb-5687, MR-1, Tahccp2 |
Location: |
Chr.1:74284597-74353692 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pnkd |